Ethyl pyruvate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Ethyl 2-oxopropanoate, 2-Oxopropionic acid ethyl ester, α-Ketopropionic acid ethyl ester, Pyruvic acid ethyl ester, 2-Ketopropionic acid ethyl ester
A1225662-25G
A1225662-100G
76679-340EA
5
USD
76679-340
77866-835
Ethyl pyruvate 95%
Ethyl pyruvate
Precaución: for Research Use Only
Formula:
C₅H₈O₃ |
MDL Number:
MFCD00009123 CAS Number: 617-35-6 UN: 3272 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...