Benzyl (cis-4-(hydroxymethyl)cyclohexyl)carbamate 97+%

Supplier: Ambeed
Warning

A1225109-1G A1225109-250MG A1225109-5G
76679-334EA 55 USD
76679-334 76679-336 76679-338
Benzyl (cis-4-(hydroxymethyl)cyclohexyl)carbamate 97+%
Benzyl (cis-4-(hydroxymethyl)cyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₃
MDL Number: MFCD26131316
CAS Number: 1256276-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR