Triphenyl(4-pyridylmethyl)phosphonium chloride 95%

Supplier: Ambeed
Warning

A1223380-100MG A1223380-1G
76679-310EA 19 USD
76679-310 76679-312
Triphenyl(4-pyridylmethyl)phosphonium chloride 95%
Triphenyl(4-pyridylmethyl)phosphonium chloride

Precaución: for Research Use Only

Formula: C₂₄H₂₁ClNP
MW: 389.8570 g/mol
MDL Number: MFCD03411793
CAS Number: 73870-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR