1-Bromo-4-(2-bromo-1,1,2,2-tetrafluoro-ethoxy)benzene 95%

Supplier: Ambeed
Warning

A1223141-100MG A1223141-1G A1223141-250MG
76679-300EA 17 USD
76679-300 76679-302 76679-304
1-Bromo-4-(2-bromo-1,1,2,2-tetrafluoro-ethoxy)benzene 95%
1-Bromo-4-(2-bromo-1,1,2,2-tetrafluoro-ethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₄Br₂F₄O
MDL Number: MFCD31546217
CAS Number: 113939-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR