6-Bromo-1-(trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline 95%

Supplier: Ambeed
Warning

A1221741-250MG A1221741-1G
76679-290EA 245.07 USD
76679-290 77596-432
6-Bromo-1-(trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline 95%
6-Bromo-1-(trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline

Precaución: for Research Use Only

Formula: C₁₀H₉BrF₃N
MW: 280.08 g/mol
MDL Number: MFCD22520970
CAS Number: 1391067-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR