Propargyl-PEG5-Tos 98%

Supplier: Ambeed
Warning

Synonyms: 3,6,9,12-Tetraoxapentadec-14-yn-1-yl 4-methylbenzenesulphonate

A1221415-100MG A1221415-1G A1221415-250MG
76679-284EA 22 USD
76679-284 76679-286 76679-288
Propargyl-PEG5-Tos 98%
Propargyl-PEG5-Tos

Precaución: for Research Use Only

Formula: C₁₈H₂₆O₇S
MDL Number: MFCD28950765
CAS Number: 875770-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR