4-(3-Bromo-5-chlorophenyl)-2,6-diphenylpyrimidine 97%

Supplier: Ambeed
Warning

A1221350-1G A1221350-250MG A1221350-5G
76679-280EA 31 USD
76679-280 76679-282 77866-806
4-(3-Bromo-5-chlorophenyl)-2,6-diphenylpyrimidine 97%
4-(3-Bromo-5-chlorophenyl)-2,6-diphenylpyrimidine

Precaución: for Research Use Only

Formula: C₂₂H₁₄BrClN₂
CAS Number: 2088827-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR