4-(Methylamino)phenol sulfate 99%
Supplier: Ambeed
|
Danger
|
A1221285-25G
A1221285-5G
A1221285-100G
76679-276EA
14
USD
76679-276
76679-278
77866-805
4-(Methylamino)phenol sulfate 99%
4-(Methylamino)phenol sulfate
Precaución: for Research Use Only
Formula:
C₁₄H₂₀N₂O₆S |
MDL Number:
MFCD00013140 CAS Number: 55-55-0 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...