Methyl 4-amino-6-chloropyrimidine-5-carboxylate 97%

Supplier: Ambeed
Danger

A1221079-100MG A1221079-250MG
76679-264EA 103 USD
76679-264 76679-266
Methyl 4-amino-6-chloropyrimidine-5-carboxylate 97%
Methyl 4-amino-6-chloropyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆ClN₃O₂
CAS Number: 2091735-99-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR