4-(4-Chlorobenzyl)-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidin-4-amine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: CCT128930 HCl

A1221053-10MG A1221053-1MG A1221053-25MG A1221053-50MG A1221053-5MG
76679-248EA 91.24 USD
76679-248 76679-250 76679-252 76679-254 76679-256
4-(4-Chlorobenzyl)-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidin-4-amine hydrochloride 98%
4-(4-Chlorobenzyl)-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidin-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₂₁Cl₂N₅
MW: 378.3 g/mol
MDL Number: MFCD32667039
CAS Number: 2453324-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR