Methyl 3-(tert-butyl)-[1,2,4]triazolo[4,3-a]pyridine-6-carboxylate 97%

Supplier: Ambeed
Warning

A1221047-100MG A1221047-1G A1221047-250MG
76679-240EA 29.71 USD
76679-240 76679-242 76679-244
Methyl 3-(tert-butyl)-[1,2,4]triazolo[4,3-a]pyridine-6-carboxylate 97%
Methyl 3-(tert-butyl)-[1,2,4]triazolo[4,3-a]pyridine-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₂
MW: 233.2664 g/mol
MDL Number: MFCD25195306
CAS Number: 1549306-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR