(S)-2-Ethylbutyl 2-(((S)-(4-nitrophenoxy)(phenoxy)phosphoryl)amino)propanoate 97%

Supplier: Ambeed
Warning

Synonyms: N-[(S)-(4-Nitrophenoxy)phenoxyphosphinyl]-L-alanine 2-ethylbutyl ester

A1220961-1G A1220961-250MG A1220961-25G A1220961-5G
76679-234EA 25 USD
76679-234 76679-236 76679-238 77571-348
(S)-2-Ethylbutyl 2-(((S)-(4-nitrophenoxy)(phenoxy)phosphoryl)amino)propanoate 97%
(S)-2-Ethylbutyl 2-(((S)-(4-nitrophenoxy)(phenoxy)phosphoryl)amino)propanoate

Precaución: for Research Use Only

Formula: C₂₁H₂₇N₂O₇P
MW: 450.4221 g/mol
MDL Number: MFCD32670644
CAS Number: 1354823-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR