(3aR,4R,6R,6aR)-4-(4-Aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-6-(hydroxymethyl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxole-4-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: Remdesivir O-desphosphate acetonide impurity

A1220957-100MG A1220957-1G A1220957-250MG A1220957-5G A1220957-25G
76679-226EA 15.92 USD
76679-226 76679-228 76679-230 76679-232 77621-108
(3aR,4R,6R,6aR)-4-(4-Aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-6-(hydroxymethyl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxole-4-carbonitrile 97%
(3aR,4R,6R,6aR)-4-(4-Aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-6-(hydroxymethyl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₁₅H₁₇N₅O₄
MW: 331.33 g/mol
MDL Number: MFCD32693253
CAS Number: 1191237-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR