Lithium chloride 99%, powder

Supplier: Ambeed
Warning

A1220651-100G A1220651-500G A1220651-1000G A1220651-25G
76679-216EA 32 USD
76679-216 76679-218 77566-296 77571-344
Lithium chloride 99%, powder
Lithium chloride

Precaución: For research use only.

Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382
Melting Pt: 614
MDL Number: MFCD00011078
CAS Number: 7447-41-8
Merck Index: 13,05550

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR