N2-(3-Methoxyphenyl)-N4-((tetrahydrofuran-2-yl)methyl)quinazoline-2,4-diamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
LCH-7749944
A1220481-10MG
A1220481-1MG
A1220481-25MG
A1220481-50MG
A1220481-5MG
76679-180EA
126.25
USD
76679-180
76679-182
76679-184
76679-186
76679-188
N2-(3-Methoxyphenyl)-N4-((tetrahydrofuran-2-yl)methyl)quinazoline-2,4-diamine 98%
N2-(3-Methoxyphenyl)-N4-((tetrahydrofuran-2-yl)methyl)quinazoline-2,4-diamine
Precaución: for Research Use Only
Formula:
C₂₀H₂₂N₄O₂ MW: 350.4143 g/mol |
MDL Number:
MFCD04218695 CAS Number: 796888-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...