3-Bromo-2-methoxy-6-phenylpyridine 98%

Supplier: Ambeed
Warning

A1220357-100MG A1220357-1G A1220357-250MG
76679-174EA 147.47 USD
76679-174 76679-176 76679-178
3-Bromo-2-methoxy-6-phenylpyridine 98%
3-Bromo-2-methoxy-6-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO
MW: 264.1179 g/mol
MDL Number: MFCD18256009
CAS Number: 1256788-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR