[(4R)-4,5-Dihydro-4-phenylmethyl-2-oxazolyl]ferrocene ≥97%, ee 99%

Supplier: Ambeed
Warning

A1220227-100MG A1220227-1G A1220227-250MG
76679-148EA 31.83 USD
76679-148 76679-150 76679-152
[(4R)-4,5-Dihydro-4-phenylmethyl-2-oxazolyl]ferrocene ≥97%, ee 99%
[(4R)-4,5-Dihydro-4-phenylmethyl-2-oxazolyl]ferrocene

Precaución: for Research Use Only

Formula: C₂₀H₁₀FeNO
CAS Number: 2757082-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR