[(4R)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]ferrocene ≥97%, ee 99%

Supplier: Ambeed
Warning

A1220226-100MG A1220226-1G A1220226-250MG A1220226-5G
76679-142EA 43 USD
76679-142 76679-144 76679-146 77866-794
[(4R)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]ferrocene ≥97%, ee 99%
[(4R)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]ferrocene

Precaución: for Research Use Only

Formula: C₁₆H₁₀FeNO
MW: 288.1017 g/mol
CAS Number: 2757082-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR