(4R,4'R)-2,2'-(Cyclopropane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1220215-1G A1220215-250MG A1220215-100MG
76679-132EA 281 USD
76679-132 76679-134 77866-792
(4R,4'R)-2,2'-(Cyclopropane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 97%
(4R,4'R)-2,2'-(Cyclopropane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₁₇H₂₈N₂O₂
CAS Number: 2634687-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR