(4S,4'S)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1220210-100MG A1220210-1G A1220210-250MG
76679-110EA 103.97 USD
76679-110 76679-112 76679-114
(4S,4'S)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 97%
(4S,4'S)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₃H₃₂N₂O₂
MW: 368.5124 g/mol
CAS Number: 2757082-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR