(4R,4'R)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 97%
Supplier: Ambeed
|
Warning
|
A1220209-100MG
A1220209-1G
A1220209-250MG
76679-104EA
150.65
USD
76679-104
76679-106
76679-108
(4R,4'R)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 97%
(4R,4'R)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole)
Precaución: for Research Use Only
Formula:
C₂₃H₃₂N₂O₂ MW: 368.5124 g/mol |
CAS Number:
2757082-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...