(4R,4'R)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-isopropyl-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1220207-100MG A1220207-1G A1220207-250MG
76679-094EA 129 USD
76679-094 76679-096 77596-424
(4R,4'R)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-isopropyl-4,5-dihydrooxazole) 97%
(4R,4'R)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-isopropyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₁H₂₈N₂O₂
MDL Number: MFCD32899508
CAS Number: 2757082-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR