(4R,4'R)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-phenyl-4,5-dihydrooxazole) ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A1220206-100MG
A1220206-1G
A1220206-250MG
76679-088EA
124.13
USD
76679-088
76679-090
76679-092
(4R,4'R)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-phenyl-4,5-dihydrooxazole) ≥97%, ee 99%
(4R,4'R)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-phenyl-4,5-dihydrooxazole)
Precaución: for Research Use Only
Formula:
C₂₇H₂₄N₂O₂ MW: 408.4917 g/mol |
CAS Number:
2757082-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...