(R)-4-(tert-Butyl)-2-(4-(trifluoromethyl)pyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1220182-100MG A1220182-1G A1220182-250MG
76679-072EA 124.13 USD
76679-072 76679-074 76679-076
(R)-4-(tert-Butyl)-2-(4-(trifluoromethyl)pyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%
(R)-4-(tert-Butyl)-2-(4-(trifluoromethyl)pyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₃H₁₅F₃N₂O
CAS Number: 2828432-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR