(4R,4'R)-2,2'-(Cyclohexane-1,1-diyl)bis(4-phenyl-4,5-dihydrooxazole) ≥97%, ee 99%

Supplier: Ambeed
Warning

A1220165-100MG A1220165-250MG A1220165-50MG A1220165-1G
76679-000EA 55 USD
76679-000 76679-002 76679-004 77866-788
(4R,4'R)-2,2'-(Cyclohexane-1,1-diyl)bis(4-phenyl-4,5-dihydrooxazole) ≥97%, ee 99%
(4R,4'R)-2,2'-(Cyclohexane-1,1-diyl)bis(4-phenyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₄H₂₆N₂O₂
MW: 374.4700 g/mol
CAS Number: 2271404-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR