(4R,4'R)-2,2'-(Cyclohexane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) ≥97%, ee 99%

Supplier: Ambeed
Warning

A1220163-100MG A1220163-1G A1220163-250MG A1220163-5G
76678-992EA 47 USD
76678-992 76678-994 76678-996 76678-998
(4R,4'R)-2,2'-(Cyclohexane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) ≥97%, ee 99%
(4R,4'R)-2,2'-(Cyclohexane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₀H₃₄N₂O₂
MW: 334.4962 g/mol
MDL Number: MFCD32201211
CAS Number: 2757082-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR