(4R,4'R)-2,2'-(Cyclobutane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1220158-100MG A1220158-1G A1220158-250MG
76678-972EA 150.65 USD
76678-972 76678-974 76678-976
(4R,4'R)-2,2'-(Cyclobutane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 97%
(4R,4'R)-2,2'-(Cyclobutane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₁₈H₃₀N₂O₂
MW: 306.4400 g/mol
CAS Number: 2828431-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR