(4R,4'R,5S,5'S)-2,2'-(Cyclobutane-1,1-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) 97%
Supplier: Ambeed
|
Warning
|
A1220155-100MG
A1220155-1G
A1220155-250MG
76678-964EA
165.51
USD
76678-964
76678-966
76678-968
(4R,4'R,5S,5'S)-2,2'-(Cyclobutane-1,1-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) 97%
(4R,4'R,5S,5'S)-2,2'-(Cyclobutane-1,1-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole)
Precaución: for Research Use Only
Formula:
C₃₄H₃₀N₂O₂ MW: 498.6100 g/mol |
CAS Number:
2828431-95-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...