Bis(4-fluorophenyl)phosphine oxide 95% (by GC)

Supplier: Ambeed
Danger

A1220057-1G A1220057-250MG A1220057-25G A1220057-5G
76678-944EA 97.61 USD
76678-944 76678-946 77621-066 77621-068
Bis(4-fluorophenyl)phosphine oxide 95% (by GC)
Bis(4-fluorophenyl)phosphine oxide

Precaución: for Research Use Only

Formula: C₁₂H₉F₂OP
MW: 238.17 g/mol
CAS Number: 94940-35-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR