Di-o-tolylphosphine oxide 97%

Supplier: Ambeed
Danger

A1220055-1G A1220055-250MG A1220055-5G A1220055-25G
76678-938EA 39 USD
76678-938 76678-940 76678-942 77866-783
Di-o-tolylphosphine oxide 97%
Di-o-tolylphosphine oxide

Precaución: for Research Use Only

Formula: C₁₄H₁₅OP
MDL Number: MFCD06754096
CAS Number: 30309-80-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR