Dimesitylphosphine oxide 97%

Supplier: Ambeed
Danger

A1220053-1G A1220053-25G
76678-928EA 31 USD
76678-928 76678-930
Dimesitylphosphine oxide 97%
Dimesitylphosphine oxide

Precaución: for Research Use Only

Formula: C₁₈H₂₃OP
MDL Number: MFCD00418340
CAS Number: 23897-16-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR