Bis(4-(tert-butyl)phenyl)phosphine oxide 97%

Supplier: Ambeed
Danger

A1220052-100MG A1220052-1G A1220052-250MG A1220052-5G
76678-922EA 52 USD
76678-922 76678-924 76678-926 77866-782
Bis(4-(tert-butyl)phenyl)phosphine oxide 97%
Bis(4-(tert-butyl)phenyl)phosphine oxide

Precaución: for Research Use Only

Formula: C₂₀H₂₇OP
MDL Number: MFCD32856151
CAS Number: 326921-37-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR