5-((3aR,8aR)-2,2-Dimethyl-4,4,8,8-tetraphenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)-5H-dibenzo[b,f]azepine 97%

Supplier: Ambeed
Warning

A1220049-100MG A1220049-250MG
76678-918EA 100.79 USD
76678-918 76678-920
5-((3aR,8aR)-2,2-Dimethyl-4,4,8,8-tetraphenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)-5H-dibenzo[b,f]azepine 97%
5-((3aR,8aR)-2,2-Dimethyl-4,4,8,8-tetraphenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)-5H-dibenzo[b,f]azepine

Precaución: for Research Use Only

Formula: C₄₅H₃₈NO₄P
MW: 687.7613 g/mol
MDL Number: MFCD22417198
CAS Number: 1092695-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR