2,6-Bis((S)-4-((S)-sec-butyl)-4,5-dihydrooxazol-2-yl)pyridine 97%

Supplier: Ambeed
Warning

A1220016-100MG A1220016-1G A1220016-250MG
76678-866EA 110 USD
76678-866 76678-868 77596-418
2,6-Bis((S)-4-((S)-sec-butyl)-4,5-dihydrooxazol-2-yl)pyridine 97%
2,6-Bis((S)-4-((S)-sec-butyl)-4,5-dihydrooxazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₉H₂₇N₃O₂
CAS Number: 118949-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR