(4R,4'R,5S,5'S)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A1220014-100MG
A1220014-1G
A1220014-250MG
76678-860EA
82
USD
76678-860
76678-862
76678-864
(4R,4'R,5S,5'S)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) ≥97%, ee 99%
(4R,4'R,5S,5'S)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole)
Precaución: for Research Use Only
Formula:
C₃₉H₃₂N₂O₂ |
CAS Number:
1656253-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...