(4S,4'S)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-phenyl-4,5-dihydrooxazole) ≥97%, ee 99%

Supplier: Ambeed
Warning

A1220013-100MG A1220013-1G A1220013-250MG
76678-854EA 112.46 USD
76678-854 76678-856 76678-858
(4S,4'S)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-phenyl-4,5-dihydrooxazole) ≥97%, ee 99%
(4S,4'S)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-phenyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₇H₂₄N₂O₂
MW: 408.4917 g/mol
CAS Number: 188780-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR