(S)-2-(2-(Bis(4-(trifluoromethyl)phenyl)phosphino)-5-(trifluoromethyl)phenyl)-4-(tert-butyl)-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1219993-250MG A1219993-50MG
76678-834EA 265.23 USD
76678-834 76678-836
(S)-2-(2-(Bis(4-(trifluoromethyl)phenyl)phosphino)-5-(trifluoromethyl)phenyl)-4-(tert-butyl)-4,5-dihydrooxazole 97%
(S)-2-(2-(Bis(4-(trifluoromethyl)phenyl)phosphino)-5-(trifluoromethyl)phenyl)-4-(tert-butyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₈H₂₃F₉NOP
MW: 591.4477 g/mol
CAS Number: 944836-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR