2-(2-(Diphenylphosphino)phenyl)-4,4-dimethyl-4,5-dihydrooxazole 97%
Supplier: Ambeed
|
Warning
|
A1219991-100MG
A1219991-250MG
76678-822EA
42.44
USD
76678-822
76678-824
2-(2-(Diphenylphosphino)phenyl)-4,4-dimethyl-4,5-dihydrooxazole 97%
2-(2-(Diphenylphosphino)phenyl)-4,4-dimethyl-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₂₃H₂₂NOP MW: 359.4007 g/mol |
CAS Number:
64691-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...