(2S)-1-[(4R)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]-2-(diphenylphosphino)ferrocene ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2Rp)-1-[(4R)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]-2-(diphenylphosphino)ferrocene
A1219990-100MG
A1219990-1G
A1219990-250MG
A1219990-5G
76678-816EA
21
USD
76678-816
76678-818
76678-820
77621-054
(2S)-1-[(4R)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]-2-(diphenylphosphino)ferrocene ≥97%, ee 99%
(2S)-1-[(4R)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]-2-(diphenylphosphino)ferrocene
Precaución: for Research Use Only
Formula:
C₂₈H₂₈FeNOP MW: 481.3600 g/mol |
CAS Number:
1065474-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...