(2R)-1-[(4S)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]-2-(diphenylphosphino)ferrocene ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A1219989-100MG
A1219989-250MG
A1219989-1G
A1219989-5G
76678-812EA
23
USD
76678-812
76678-814
77592-376
77596-416
(2R)-1-[(4S)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]-2-(diphenylphosphino)ferrocene ≥97%, ee 99%
(2R)-1-[(4S)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]-2-(diphenylphosphino)ferrocene
Precaución: for Research Use Only
Formula:
C₂₈H₂₈FeNOP MW: 481.3600 g/mol |
MDL Number:
MFCD15144869 CAS Number: 163169-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...