(S)-(5-tert-Butyloxazolidinyl)ferrocene 97%

Supplier: Ambeed
Warning

A1219947-100MG A1219947-1G A1219947-250MG
76678-766EA 15 USD
76678-766 76678-768 76678-770
(S)-(5-tert-Butyloxazolidinyl)ferrocene 97%
(S)-(5-tert-Butyloxazolidinyl)ferrocene

Precaución: for Research Use Only

Formula: C₁₇H₁₂FeNO
CAS Number: 162157-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR