(S)-4-Phenyl-2-(5-(trifluoromethyl)pyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1219934-100MG A1219934-250MG A1219934-1G
76678-696EA 47 USD
76678-696 76678-698 77621-032
(S)-4-Phenyl-2-(5-(trifluoromethyl)pyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%
(S)-4-Phenyl-2-(5-(trifluoromethyl)pyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃N₂O
MW: 292.2559 g/mol
MDL Number: MFCD32173539
CAS Number: 1803416-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR