(4R,4'R,5S,5'S)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) ≥97%, ee 99%

Supplier: Ambeed
Warning

A1219931-100MG A1219931-1G A1219931-250MG
76678-680EA 51 USD
76678-680 76678-682 76678-684
(4R,4'R,5S,5'S)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) ≥97%, ee 99%
(4R,4'R,5S,5'S)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₅₃H₅₄N₂O₂
CAS Number: 1908437-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR