(4S,4'S)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-isopropyl-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1219930-1G A1219930-250MG
76678-676EA 315 USD
76678-676 76678-678
(4S,4'S)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-isopropyl-4,5-dihydrooxazole) 97%

Precaución: for Research Use Only

Formula: C₃₅H₅₀N₂O₂
MDL Number: MFCD32671436
CAS Number: 1373357-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR