(4S,4'S)-2,2'-(Cyclohexane-1,1-diyl)bis(4-benzyl-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1219921-100MG A1219921-1G A1219921-250MG
76678-646EA 90.18 USD
76678-646 76678-648 76678-650
(4S,4'S)-2,2'-(Cyclohexane-1,1-diyl)bis(4-benzyl-4,5-dihydrooxazole) 97%
(4S,4'S)-2,2'-(Cyclohexane-1,1-diyl)bis(4-benzyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₆H₃₀N₂O₂
MW: 402.5286 g/mol
MDL Number: MFCD32639830
CAS Number: 1003886-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR