(4S,4'S)-2,2'-(Cyclobutane-1,1-diyl)bis(4-benzyl-4,5-dihydrooxazole) 95%

Supplier: Ambeed
Warning

A1219917-1G A1219917-250MG
76678-630EA 734.15 USD
76678-630 76678-632
(4S,4'S)-2,2'-(Cyclobutane-1,1-diyl)bis(4-benzyl-4,5-dihydrooxazole) 95%
(4S,4'S)-2,2'-(Cyclobutane-1,1-diyl)bis(4-benzyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₄H₂₆N₂O₂
MW: 374.4755 g/mol
MDL Number: MFCD32670845
CAS Number: 1003886-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR