(3aR,3a'R,8aS,8a'S)-2,2'-(Cyclobutane-1,1-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole) ≥97%, ee 99%

Supplier: Ambeed
Warning

A1219915-100MG A1219915-1G A1219915-250MG
76678-620EA 90.18 USD
76678-620 76678-622 76678-624
(3aR,3a'R,8aS,8a'S)-2,2'-(Cyclobutane-1,1-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole) ≥97%, ee 99%
(3aR,3a'R,8aS,8a'S)-2,2'-(Cyclobutane-1,1-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole)

Precaución: for Research Use Only

Formula: C₂₄H₂₂N₂O₂
MW: 370.4400 g/mol
CAS Number: 2097145-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR