(4S,4'S)-2,2'-(Cyclopropane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 95%

Supplier: Ambeed
Warning

A1219914-250MG A1219914-100MG A1219914-1G
76678-618EA 89 USD
76678-618 77596-410 77621-024
(4S,4'S)-2,2'-(Cyclopropane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 95%
(4S,4'S)-2,2'-(Cyclopropane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₁₇H₂₈N₂O₂
CAS Number: 195379-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR