(4R,4'R,5S,5'S)-2,2'-(Cyclopropane-1,1-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) 97%
Supplier: Ambeed
|
Warning
|
A1219911-100MG
A1219911-1G
A1219911-250MG
76678-606EA
32
USD
76678-606
76678-608
76678-610
(4R,4'R,5S,5'S)-2,2'-(Cyclopropane-1,1-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) 97%
(4R,4'R,5S,5'S)-2,2'-(Cyclopropane-1,1-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole)
Precaución: for Research Use Only
Formula:
C₃₃H₂₈N₂O₂ |
CAS Number:
229184-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...