(4S,4'S)-4,4'-Bis(1,1-dimethylethyl)-4,4',5,5'-tetrahydro-2,2'-bioxazole 97%

Supplier: Ambeed
Warning

A1219905-100MG A1219905-1G A1219905-250MG A1219905-5G
76678-590EA 40.32 USD
76678-590 77566-282 77566-284 77621-022
(4S,4'S)-4,4'-Bis(1,1-dimethylethyl)-4,4',5,5'-tetrahydro-2,2'-bioxazole 97%
(4S,4'S)-4,4'-Bis(1,1-dimethylethyl)-4,4',5,5'-tetrahydro-2,2'-bioxazole

Precaución: For research use only.

Formula: C₁₄H₂₄N₂O₂
MW: 252.3526 g/mol
MDL Number: MFCD32671514
CAS Number: 135565-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR